The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015537.4 | NP_056352.3 | 4 - 5 | 934 | 86 |
XM_005266061.4 | XP_005266118.1 | 4 - 5 | 957 | 86 | |
GenBank mRNA | AK027474.1 | - | 2 - 3 | 205 | 86 |
AK074602.1 | - | 4 - 5 | 902 | 86 | |
AY255129.1 | - | 4 - 5 | 934 | 86 | |
BC072412.1 | - | 4 - 5 | 919 | 86 |