The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016848.5 | NP_058544.3 | 4 - 5 | 1041 | 70 |
XM_011518785.2 | XP_011517087.1 | 4 - 5 | 4571 | 70 | |
XM_011518786.2 | XP_011517088.1 | 5 - 6 | 420 | 70 | |
XM_017014810.1 | XP_016870299.1 | 4 - 5 | 4582 | 70 | |
GenBank mRNA | BC026314.1 | - | 4 - 5 | 1042 | 70 |
D84361.1 | - | 4 - 5 | 914 | 70 | |
KU176125.1 | - | 4 - 5 | 338 | 70 |