The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017952.5 | NP_060422.4 | 17 - 18 | 1442 | 105 |
GenBank mRNA | AK000765.1 | - | 7 - 8 | 613 | 105 |
AK315676.1 | - | 11 - 12 | 896 | 105 | |
AL832520.1 | - | 6 - 7 | 446 | 105 | |
AY277598.1 | - | 5 - 6 | 410 | 105 | |
BC001758.1 | - | 6 - 7 | 516 | 105 | |
BC011832.2 | - | 17 - 18 | 1377 | 105 |