The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020433.4 | NP_065166.2 | 3 - 4 | 2160 | 54 |
GenBank mRNA | AK304863.1 | - | 3 - 4 | 805 | 54 |
BC043206.2 | - | 2 - 3 | 450 | 54 |