The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199173.1 | NP_001186102.1 | 4 - 5 | 709 | 83 |
NM_001199174.1 | NP_001186103.1 | 4 - 5 | 548 | 83 | |
NM_001199175.1 | NP_001186104.1 | 4 - 5 | 736 | 83 | |
NM_022372.4 | NP_071767.3 | 4 - 5 | 739 | 83 | |
XM_005255475.3 | XP_005255532.2 | 4 - 5 | 515 | 83 | |
XM_005255478.2 | XP_005255535.1 | 4 - 5 | 720 | 83 | |
XM_005255479.2 | XP_005255536.1 | 4 - 5 | 647 | 83 | |
XM_017023548.1 | XP_016879037.1 | 4 - 5 | 704 | 83 | |
XM_017023549.1 | XP_016879038.1 | 4 - 5 | 445 | 83 | |
GenBank mRNA | AF195883.1 | - | 1 - 2 | 60 | 83 |
AK021536.1 | - | 4 - 5 | 708 | 83 | |
AK022227.1 | - | 4 - 5 | 518 | 83 | |
AK098762.1 | - | 4 - 5 | 644 | 83 | |
AK302201.1 | - | 3 - 4 | 548 | 83 | |
AU121635.1 | - | 4 - 5 | 518 | 83 | |
AY223837.1 | - | 3 - 4 | 549 | 83 | |
BC001313.2 | - | 4 - 5 | 429 | 83 | |
BC017119.1 | - | 3 - 4 | 440 | 83 | |
BC052292.1 | - | 3 - 4 | 963 | 83 | |
BC088354.1 | - | 4 - 5 | 457 | 83 | |
BI761765.1 | - | 4 - 5 | 467 | 83 |