Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS3
Entrez Gene ID
64785
Gene Name
GINS complex subunit 3
Gene Aliases
PSF3
Chromosome Location
Chr.16: 58392394 - 58406144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022770.3 NP_073607.2 1 - 2 394 60
GenBank mRNA AK315622.1 - 1 - 2 345 60
BC005879.1 - 1 - 2 269 60
BC014437.1 - 1 - 2 288 60
DA550725.1 - 1 - 2 394 60
KU178752.1 - 1 - 2 186 60

View Full Product Details