The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024052.4 | NP_076957.3 | 2 - 3 | 671 | 115 |
XM_005256799.4 | XP_005256856.1 | 2 - 3 | 558 | 115 | |
XM_005256800.1 | XP_005256857.1 | 2 - 3 | 558 | 115 | |
XM_011524008.1 | XP_011522310.1 | 2 - 3 | 558 | 115 | |
GenBank mRNA | AK074284.1 | - | 2 - 3 | 546 | 115 |
AK127580.1 | - | 2 - 3 | 358 | 115 | |
AK315083.1 | - | 2 - 3 | 558 | 116 | |
BC000636.2 | - | 2 - 3 | 332 | 115 | |
BC041829.1 | - | 2 - 3 | 676 | 115 |