The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024052.4 | NP_076957.3 | 4 - 5 | 875 | 84 |
GenBank mRNA | AK074284.1 | - | 4 - 5 | 750 | 84 |
AK127580.1 | - | 4 - 5 | 562 | 84 | |
AK315083.1 | - | 4 - 5 | 763 | 84 | |
BC000636.2 | - | 4 - 5 | 536 | 84 | |
BC041829.1 | - | 4 - 5 | 880 | 84 |