Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
79663
Gene Name
HSPB1 associated protein 1
Gene Aliases
PASS1
Chromosome Location
Chr.3: 122739997 - 122793829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024610.5 NP_078886.2 5 - 6 878 77
XM_011513130.2 XP_011511432.1 6 - 7 796 77
XM_011513132.2 XP_011511434.2 5 - 6 637 77
XM_017007179.1 XP_016862668.1 4 - 5 702 77
GenBank mRNA AF400663.1 - 5 - 6 845 77
BC011897.2 - 5 - 6 862 77

View Full Product Details