Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO31
Entrez Gene ID
79791
Gene Name
F-box protein 31
Gene Aliases
FBX14, FBXO14, Fbx31, MRT45, pp2386
Chromosome Location
Chr.16: 87326987 - 87392107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282683.1 NP_001269612.1 6 - 7 528 101
NM_024735.4 NP_079011.3 5 - 6 780 101
GenBank mRNA AF318348.1 - 4 - 5 980 101
AF428140.1 - 5 - 6 744 101
AL117444.1 - 3 - 4 288 101
AY736035.1 - 6 - 7 731 101
BC012748.1 - 5 - 6 754 101
BG718342.1 - 6 - 7 527 101

View Full Product Details