The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032596.3 | NP_115985.2 | 3 - 4 | 617 | 74 |
XM_005251616.4 | XP_005251673.1 | 3 - 4 | 3599 | 74 | |
GenBank mRNA | AK058087.1 | - | 3 - 4 | 555 | 74 |
BC022084.1 | - | 3 - 4 | 602 | 74 | |
BC029484.1 | - | 3 - 4 | 617 | 74 |