Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1072740 | Chr17:45970433 - 46248034 on Build GRCh38 | Deletion | STH MAPT KANSL1-AS1 KANSL1 |
esv2751686 | Chr17:45962198 - 46286690 on Build GRCh38 | Gain | STH LRRC37A MAPT KANSL1-AS1 ARL17B KANSL1 |
esv2758692 | Chr17:46006548 - 47199967 on Build GRCh38 | Gain+Loss | ARL17A MIR5089 NSF LOC107987253 RPS2P47 RPRML LRRC37A GOSR2 CDC27 LRRC37A2 KANSL1 NSFP1 WNT9B WNT3 LOC107987254 MAPT KANSL1-AS1 ARL17B |
dgv128e55 | Chr17:46017105 - 46286690 on Build GRCh38 | Gain | LRRC37A MAPT KANSL1-AS1 ARL17B KANSL1 |
nsv471698 | Chr17:46002113 - 46300887 on Build GRCh38 | Gain+Loss | LRRC37A MAPT KANSL1-AS1 ARL17B KANSL1 |
nsv469705 | Chr17:45969304 - 46127121 on Build GRCh38 | Loss | STH MAPT KANSL1 |
nsv469537 | Chr17:46002112 - 46163353 on Build GRCh38 | Gain+Loss | MAPT KANSL1 |
nsv2069 | Chr17:46034385 - 46065659 on Build GRCh38 | Insertion | KANSL1 |
nsv575142 | Chr17:46003698 - 46172742 on Build GRCh38 | Loss | MAPT KANSL1 |
esv2762433 | Chr17:45496768 - 46710828 on Build GRCh38 | Gain+Loss | ARL17A SPPL2C LRRC37A4P NSF LOC107987253 LOC644172 MAPT-IT1 MGC57346 LRRC37A CRHR1-IT1 LRRC37A2 KANSL1 MGC57346-CRHR1 NSFP1 MAPT-AS1 LOC105376839 CRHR1 STH LOC107987254 MAPT KANSL1-AS1 ARL17B |
dgv366n27 | Chr17:46032477 - 46215654 on Build GRCh38 | Gain | KANSL1-AS1 KANSL1 |
dgv5618n54 | Chr17:46032108 - 46330700 on Build GRCh38 | Gain | LRRC37A KANSL1-AS1 ARL17B KANSL1 |
dgv127e55 | Chr17:45927606 - 46733750 on Build GRCh38 | Gain | NSFP1 ARL17A NSF LOC107987253 STH LOC107987254 LRRC37A MAPT KANSL1-AS1 LRRC37A2 ARL17B KANSL1 |