Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
114571
Gene Name
solute carrier family 22 member 9
Gene Aliases
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location
Chr.11: 63368871 - 63410878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080866.2 NP_543142.2 5 - 6 1225 78
XM_017017159.1 XP_016872648.1 5 - 6 2143 78
XM_017017160.1 XP_016872649.1 4 - 5 802 78
GenBank mRNA AB062418.1 - 5 - 6 1058 78
AB074812.1 - 5 - 6 1206 78
AF440402.1 - 5 - 6 1193 78
AJ295270.1 - 5 - 6 1017 78
AK074246.1 - 5 - 6 1225 78
BC022379.1 - 4 - 5 1070 78
BC126288.1 - 5 - 6 1107 78

View Full Product Details