Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL2
Entrez Gene ID
9885
Gene Name
oxysterol binding protein like 2
Gene Aliases
DFNA67, DNFA67, ORP-2, ORP2
Chromosome Location
Chr.20: 62238485 - 62296213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278649.1 NP_001265578.1 6 - 7 681 76
NM_014835.3 NP_055650.1 6 - 7 700 76
NM_144498.2 NP_653081.1 6 - 7 736 76
XM_017028163.1 XP_016883652.1 6 - 7 736 76
XM_017028164.1 XP_016883653.1 7 - 8 1041 76
XM_017028165.1 XP_016883654.1 7 - 8 1041 76
XM_017028166.1 XP_016883655.1 7 - 8 790 76
XM_017028167.1 XP_016883656.1 7 - 8 804 76
XM_017028168.1 XP_016883657.1 6 - 7 736 76
XM_017028169.1 XP_016883658.1 6 - 7 660 76
XM_017028170.1 XP_016883659.1 6 - 7 660 76
GenBank mRNA AB018315.1 - 6 - 7 606 76
AF392447.1 - 6 - 7 697 76
AK000230.1 - 6 - 7 697 76
AK291851.1 - 6 - 7 664 76
AK296595.1 - 6 - 7 642 76
AK315576.1 - 6 - 7 697 76
AY028168.1 - 6 - 7 606 76
BC000296.2 - 6 - 7 640 76
BC004455.1 - 6 - 7 640 76
BC018812.2 - 6 - 7 640 76
CR456722.1 - 5 - 6 495 76

View Full Product Details