Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFXN5
Entrez Gene ID
94097
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
Chromosome Location
Chr.2: 72942036 - 73072234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144579.2 NP_653180.1 3 - 4 377 99
XM_005264648.1 XP_005264705.1 3 - 4 283 99
XM_006712147.1 XP_006712210.1 3 - 4 283 99
XM_011533186.2 XP_011531488.1 4 - 5 1925 99
XM_011533187.2 XP_011531489.1 4 - 5 1922 99
XM_011533188.2 XP_011531490.1 3 - 4 1780 99
XM_011533189.1 XP_011531491.1 3 - 4 283 99
XM_011533190.2 XP_011531492.1 4 - 5 830 99
XM_011533192.2 XP_011531494.1 4 - 5 1064 99
XM_011533194.2 XP_011531496.1 4 - 5 1923 99
XM_011533195.2 XP_011531497.1 4 - 5 1925 99
XM_011533196.2 XP_011531498.1 3 - 4 1009 99
XM_011533197.1 XP_011531499.1 3 - 4 283 99
XM_017005340.1 XP_016860829.1 3 - 4 1777 99
XM_017005341.1 XP_016860830.1 3 - 4 1778 99
XM_017005342.1 XP_016860831.1 3 - 4 283 99
XM_017005343.1 XP_016860832.1 4 - 5 1064 99
XM_017005344.1 XP_016860833.1 3 - 4 1009 99
GenBank mRNA AK024193.1 - 2 - 3 905 99
AK289731.1 - 3 - 4 266 99
AK294751.1 - 3 - 4 314 99
AK295637.1 - 3 - 4 188 99
AK314770.1 - 3 - 4 265 99
AY044437.1 - 3 - 4 381 99
BC013233.1 - 3 - 4 318 99
BC101311.2 - 3 - 4 377 99
BC101312.1 - 3 - 4 377 99
BC101313.2 - 3 - 4 377 99

View Full Product Details