The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304499.1 | NP_001291428.1 | 5 - 6 | 581 | 123 |
NM_001304500.1 | NP_001291429.1 | 3 - 4 | 311 | 123 | |
NM_144600.3 | NP_653201.1 | 4 - 5 | 509 | 123 | |
NR_130755.1 | 3 - 4 | 424 | 123 | ||
NR_130756.1 | 5 - 6 | 609 | 123 | ||
NR_130757.1 | 2 - 3 | 226 | 123 | ||
GenBank mRNA | AK055715.1 | - | 4 - 5 | 452 | 123 |
AL832498.1 | - | 4 - 5 | 472 | 123 | |
AY507846.1 | - | 4 - 5 | 465 | 123 | |
BC022321.1 | - | 4 - 5 | 460 | 123 | |
BI460301.1 | - | 5 - 6 | 547 | 123 | |
BI550657.1 | - | 3 - 4 | 400 | 123 | |
BU661163.1 | - | 2 - 3 | 177 | 123 | |
CB991926.1 | - | 5 - 6 | 598 | 123 | |
DB103294.1 | - | 3 - 4 | 255 | 123 |