Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
-
Chromosome Location
Chr.9: 130265742 - 130434123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291815.1 NP_001278744.1 97 - 98 14894 111
XM_011518465.2 XP_011516767.1 96 - 97 14908 111
XM_011518466.2 XP_011516768.1 96 - 97 14899 111
XM_011518467.2 XP_011516769.1 96 - 97 14854 111
XM_017014585.1 XP_016870074.1 76 - 77 11700 111
XM_017014586.1 XP_016870075.1 51 - 52 7628 111
GenBank mRNA AK074396.1 - 3 - 4 571 111

View Full Product Details