The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178003.1 | NP_001171474.1 | 4 - 5 | 1061 | 85 |
NM_144668.5 | NP_653269.3 | 4 - 5 | 1048 | 85 | |
GenBank mRNA | AA853421.1 | - | 1 - 2 | 73 | 85 |
AL833930.1 | - | 2 - 3 | 336 | 85 | |
BC028421.2 | - | 4 - 5 | 1046 | 85 | |
BC036233.1 | - | 4 - 5 | 1041 | 85 |