The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109763.1 | NP_001103233.1 | 6 - 7 | 986 | 55 |
NM_001323900.1 | NP_001310829.1 | 7 - 8 | 1040 | 55 | |
NM_001323901.1 | NP_001310830.1 | 5 - 6 | 833 | 55 | |
NM_144675.2 | NP_653276.1 | 5 - 6 | 602 | 55 | |
GenBank mRNA | AK128775.1 | - | 6 - 7 | 607 | 55 |
AY302134.1 | - | 5 - 6 | 490 | 55 | |
AY358206.1 | - | 5 - 6 | 825 | 55 | |
BC016460.1 | - | 5 - 6 | 602 | 55 |