The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277126.1 | NP_001264055.1 | 2 - 3 | 593 | 79 |
NM_001277129.1 | NP_001264058.1 | 2 - 3 | 593 | 79 | |
NM_144687.3 | NP_653288.1 | 2 - 3 | 593 | 79 | |
XM_011527479.1 | XP_011525781.1 | 2 - 3 | 593 | 79 | |
XM_011527480.1 | XP_011525782.1 | 2 - 3 | 593 | 79 | |
XM_011527482.1 | XP_011525784.1 | 2 - 3 | 593 | 79 | |
XM_017027460.1 | XP_016882949.1 | 2 - 3 | 593 | 79 | |
XM_017027461.1 | XP_016882950.1 | 2 - 3 | 593 | 79 | |
XM_017027462.1 | XP_016882951.1 | 2 - 3 | 593 | 79 | |
XM_017027463.1 | XP_016882952.1 | 3 - 4 | 366 | 79 | |
XM_017027464.1 | XP_016882953.1 | 2 - 3 | 164 | 79 | |
XM_017027465.1 | XP_016882954.1 | 3 - 4 | 374 | 79 | |
XM_017027466.1 | XP_016882955.1 | 3 - 4 | 371 | 79 | |
XM_017027467.1 | XP_016882956.1 | 2 - 3 | 166 | 79 | |
GenBank mRNA | AK095460.1 | - | 2 - 3 | 508 | 79 |
AK290772.1 | - | 2 - 3 | 375 | 79 | |
AY095146.1 | - | 2 - 3 | 540 | 79 | |
AY116204.1 | - | 2 - 3 | 584 | 79 | |
AY116205.1 | - | 2 - 3 | 584 | 79 | |
AY116206.1 | - | 2 - 3 | 584 | 79 | |
AY116207.1 | - | 2 - 3 | 584 | 79 | |
AY154467.1 | - | 2 - 3 | 540 | 79 | |
BC028069.1 | - | 2 - 3 | 483 | 79 |