Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
Chromosome Location
Chr.3: 19879472 - 19947036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144715.3 NP_653316.3 12 - 13 2529 95
XM_005264889.1 XP_005264946.1 12 - 13 2507 95
XM_011533382.1 XP_011531684.1 11 - 12 2466 95
XM_011533383.2 XP_011531685.2 12 - 13 1637 95
XM_011533385.2 XP_011531687.1 13 - 14 1905 95
XM_017005741.1 XP_016861230.1 14 - 15 2089 95
XM_017005743.1 XP_016861232.1 13 - 14 1856 95
XM_017005744.1 XP_016861233.1 12 - 13 1768 95
XM_017005745.1 XP_016861234.1 13 - 14 1809 95
GenBank mRNA AK057929.1 - 3 - 4 543 95
AK122616.1 - 14 - 15 2065 95
BC028198.1 - 12 - 13 2495 95

View Full Product Details