The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144717.3 | NP_653318.2 | 5 - 6 | 927 | 67 |
XM_006713665.3 | XP_006713728.1 | 5 - 6 | 1656 | 67 | |
XM_011512910.2 | XP_011511212.1 | 6 - 7 | 831 | 67 | |
XM_011512911.2 | XP_011511213.1 | 5 - 6 | 784 | 67 | |
XM_017006632.1 | XP_016862121.1 | 4 - 5 | 740 | 67 | |
GenBank mRNA | AK296832.1 | - | 5 - 6 | 799 | 67 |
AK316084.1 | - | 5 - 6 | 807 | 67 | |
AY358305.1 | - | 5 - 6 | 927 | 67 | |
BC027449.1 | - | 5 - 6 | 673 | 67 | |
BC063696.1 | - | 5 - 6 | 791 | 67 |