Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC16
Entrez Gene ID
158248
Gene Name
tetratricopeptide repeat domain 16
Gene Aliases
-
Chromosome Location
Chr.9: 127716064 - 127731600 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144965.2 NP_659402.1 3 - 4 364 99
XM_005251733.1 XP_005251790.1 2 - 3 176 99
XM_005251734.1 XP_005251791.1 3 - 4 349 99
XM_006716970.1 XP_006717033.1 3 - 4 349 99
XM_006716971.1 XP_006717034.1 2 - 3 176 99
XM_011518283.1 XP_011516585.1 3 - 4 362 99
XM_011518285.1 XP_011516587.1 3 - 4 362 99
XM_011518291.1 XP_011516593.1 3 - 4 349 99
XM_011518293.1 XP_011516595.1 3 - 4 362 99
XM_011518294.2 XP_011516596.1 3 - 4 362 99
XM_011518295.1 XP_011516597.1 3 - 4 362 99
XM_011518297.1 XP_011516599.1 3 - 4 362 99
XM_011518298.2 XP_011516600.1 3 - 4 362 99
GenBank mRNA AK057342.1 - 3 - 4 349 99
AK302422.1 - 2 - 3 176 99
BC031281.1 - 3 - 4 362 99

View Full Product Details