The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145011.2 | NP_659448.1 | 3 - 4 | 244 | 124 |
XM_005252384.2 | XP_005252441.1 | 4 - 5 | 482 | 124 | |
XM_005252385.2 | XP_005252442.1 | 4 - 5 | 442 | 124 | |
XM_005252386.2 | XP_005252443.1 | 3 - 4 | 789 | 124 | |
XM_011519352.2 | XP_011517654.1 | 4 - 5 | 416 | 124 | |
XM_011519353.2 | XP_011517655.1 | 5 - 6 | 498 | 124 | |
XM_011519354.2 | XP_011517656.1 | 5 - 6 | 797 | 124 | |
XM_011519355.2 | XP_011517657.1 | 4 - 5 | 853 | 124 | |
XM_011519356.2 | XP_011517658.1 | 5 - 6 | 595 | 124 | |
XM_017015837.1 | XP_016871326.1 | 4 - 5 | 812 | 124 | |
XM_017015838.1 | XP_016871327.1 | 4 - 5 | 1026 | 124 | |
GenBank mRNA | AJ491696.1 | - | 3 - 4 | 244 | 124 |
AK056452.1 | - | 3 - 4 | 352 | 124 | |
AK122958.1 | - | 4 - 5 | 339 | 124 | |
BC036038.1 | - | 4 - 5 | 369 | 124 |