The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145046.4 | NP_659483.2 | 1 - 2 | 148 | 91 |
GenBank mRNA | AB576176.1 | - | 1 - 2 | 85 | 91 |
AK058084.1 | - | 1 - 2 | 140 | 91 | |
BC014595.2 | - | 1 - 2 | 148 | 91 | |
DB459545.1 | - | 1 - 2 | 150 | 91 |