Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA5
Entrez Gene ID
166378
Gene Name
spermatogenesis associated 5
Gene Aliases
AFG2, EHLMRS, SPAF
Chromosome Location
Chr.4: 122923070 - 123319450 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317799.1 NP_001304728.1 3 - 4 516 82
NM_145207.2 NP_660208.2 3 - 4 519 82
XM_005262783.4 XP_005262840.1 3 - 4 516 82
XM_011531678.2 XP_011529980.1 3 - 4 516 82
XM_011531679.2 XP_011529981.1 3 - 4 474 82
XM_017007825.1 XP_016863314.1 3 - 4 515 82
XM_017007826.1 XP_016863315.1 3 - 4 515 82
XM_017007827.1 XP_016863316.1 3 - 4 515 82
XM_017007828.1 XP_016863317.1 3 - 4 481 82
XM_017007830.1 XP_016863319.1 3 - 4 515 82
GenBank mRNA AF361489.1 - 3 - 4 515 82
AF479656.1 - 3 - 4 489 82
AK091384.1 - 3 - 4 474 82
AL832175.1 - 3 - 4 512 82
BC048217.1 - 3 - 4 459 82
DA619323.1 - 3 - 4 517 82

View Full Product Details