Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FANK1
- Entrez Gene ID
-
92565
- Gene Name
- fibronectin type III and ankyrin repeat domains 1
- Gene Aliases
-
HSD13
- Chromosome Location
-
Chr.10: 125896539 - 126009599 on Build GRCh38
View Full Product Details