Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3GNT7
Entrez Gene ID
93010
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases
beta3GnT7
Chromosome Location
Chr.2: 231395624 - 231401164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145236.2 NP_660279.1 1 - 2 192 50
GenBank mRNA AF502430.1 - 1 - 2 192 50
AK126207.1 - 1 - 2 88 50

View Full Product Details