The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145291.3 | NP_660334.3 | 3 - 4 | 1431 | 86 |
XM_005247951.4 | XP_005248008.1 | 3 - 4 | 1431 | 86 | |
XM_006713864.3 | XP_006713927.1 | 3 - 4 | 1362 | 86 | |
GenBank mRNA | AK127560.1 | - | 3 - 4 | 1377 | 86 |
AK292551.1 | - | 3 - 4 | 1431 | 86 | |
BC016477.1 | - | 1 - 2 | 1105 | 86 | |
BC109087.1 | - | 3 - 4 | 1377 | 86 |