The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145805.2 | NP_665804.1 | 1 - 2 | 216 | 64 |
XM_017022505.1 | XP_016877994.1 | 1 - 2 | 216 | 64 | |
GenBank mRNA | AB593133.1 | - | 1 - 2 | 139 | 64 |
AK001022.1 | - | 1 - 2 | 135 | 64 | |
BC011967.1 | - | 1 - 2 | 122 | 64 | |
BC012136.1 | - | 1 - 2 | 134 | 64 | |
DB055557.1 | - | 1 - 2 | 218 | 64 |