Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIFM1
Entrez Gene ID
9131
Gene Name
apoptosis inducing factor, mitochondria associated 1
Gene Aliases
AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8
Chromosome Location
Chr.X: 130129362 - 130165887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130846.3 NP_001124318.2 2 - 3 1055 73
NM_001130847.3 NP_001124319.1 12 - 13 1601 73
NM_004208.3 NP_004199.1 11 - 12 1398 73
NM_145812.2 NP_665811.1 11 - 12 1386 73
NR_132647.1 11 - 12 1458 73
GenBank mRNA AF100928.1 - 11 - 12 1209 73
AF131759.1 - 8 - 9 750 73
AK000775.1 - 11 - 12 1234 73
AK314446.1 - 11 - 12 1186 73
AL049703.1 - 11 - 12 1251 73
AL049704.1 - 3 - 4 330 73
BC111065.1 - 11 - 12 1329 73
BC139738.1 - 11 - 12 1302 73
CR457379.1 - 11 - 12 1167 73
DQ016496.2 - 2 - 3 1055 73
DQ016498.1 - 12 - 13 1378 73
DQ016500.1 - 11 - 12 1235 73

View Full Product Details