The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 7 - 8 | 1359 | 107 |
NM_147127.4 | NP_667338.3 | 7 - 8 | 921 | 107 | |
XM_017007735.1 | XP_016863224.1 | 7 - 8 | 932 | 107 | |
XM_017007736.1 | XP_016863225.1 | 8 - 9 | 910 | 107 | |
XM_017007737.1 | XP_016863226.1 | 8 - 9 | 797 | 107 | |
XM_017007738.1 | XP_016863227.1 | 7 - 8 | 932 | 107 | |
XM_017007739.1 | XP_016863228.1 | 8 - 9 | 963 | 107 | |
GenBank mRNA | AB083067.1 | - | 6 - 7 | 627 | 107 |
AY152402.1 | - | 7 - 8 | 1356 | 107 | |
AY152403.1 | - | 7 - 8 | 1356 | 107 | |
AY185210.1 | - | 7 - 8 | 921 | 107 | |
BC143836.1 | - | 8 - 9 | 968 | 107 |