Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAT1
Entrez Gene ID
9
Gene Name
N-acetyltransferase 1
Gene Aliases
AAC1, MNAT, NAT-1, NATI
Chromosome Location
Chr.8: 18170419 - 18223689 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160171.3 NP_001153643.1 1 - 2 75 113
XM_011544688.1 XP_011542990.1 1 - 2 86 113
GenBank mRNA BC013732.1 - 1 - 2 48 113

View Full Product Details