The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031212.3 | NP_112489.3 | 1 - 2 | 414 | 65 |
XM_006718006.3 | XP_006718069.1 | 1 - 2 | 409 | 65 | |
XM_011540244.2 | XP_011538546.1 | 1 - 2 | 409 | 65 | |
GenBank mRNA | AF327402.1 | - | 1 - 2 | 314 | 65 |
AF327403.1 | - | 1 - 2 | 293 | 65 | |
AJ303077.1 | - | 1 - 2 | 314 | 65 | |
AJ303078.1 | - | 1 - 2 | 293 | 65 | |
BC047312.1 | - | 1 - 2 | 361 | 65 | |
BC058937.1 | - | 1 - 2 | 323 | 65 | |
BC076399.1 | - | 1 - 2 | 354 | 65 | |
BC094821.1 | - | 1 - 2 | 97 | 65 | |
CD672723.1 | - | 1 - 2 | 414 | 65 |