The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014023.3 | NP_054742.2 | 4 - 5 | 505 | 83 |
GenBank mRNA | AB023199.1 | - | 4 - 5 | 476 | 83 |
AK128194.1 | - | 3 - 4 | 3314 | 83 | |
AK292591.1 | - | 4 - 5 | 468 | 83 | |
AK307259.1 | - | 3 - 4 | 398 | 83 | |
AL136827.1 | - | 4 - 5 | 483 | 83 | |
BC018044.1 | - | 4 - 5 | 534 | 83 |