The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022092.2 | NP_071375.1 | 18 - 19 | 2524 | 61 |
XM_005255470.1 | XP_005255527.1 | 18 - 19 | 2503 | 61 | |
XM_005255471.3 | XP_005255528.1 | 17 - 18 | 2606 | 61 | |
XM_011522572.1 | XP_011520874.1 | 18 - 19 | 2523 | 61 | |
XM_011522573.1 | XP_011520875.1 | 10 - 11 | 1483 | 61 | |
XM_017023532.1 | XP_016879021.1 | 18 - 19 | 2487 | 61 | |
XM_017023533.1 | XP_016879022.1 | 17 - 18 | 2298 | 61 | |
XM_017023534.1 | XP_016879023.1 | 18 - 19 | 2515 | 61 | |
GenBank mRNA | AK024476.1 | - | 15 - 16 | 3930 | 61 |
AK128869.1 | - | 14 - 15 | 3533 | 61 | |
BC006278.2 | - | 18 - 19 | 2491 | 61 | |
BC006437.1 | - | 17 - 18 | 2584 | 61 | |
BC018184.1 | - | 18 - 19 | 2499 | 61 |