The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152482.2 | NP_689695.2 | 2 - 3 | 198 | 65 |
XM_005259506.3 | XP_005259563.1 | 2 - 3 | 214 | 65 | |
XM_006722653.3 | XP_006722716.1 | 2 - 3 | 173 | 65 | |
XM_017026374.1 | XP_016881863.1 | 2 - 3 | 521 | 65 | |
GenBank mRNA | AK075267.1 | - | 2 - 3 | 181 | 65 |
AK093985.1 | - | 2 - 3 | 163 | 65 | |
AL122089.1 | - | 2 - 3 | 169 | 65 | |
AY007120.1 | - | 1 - 2 | 134 | 65 | |
BC018441.1 | - | 2 - 3 | 168 | 65 | |
BP210473.1 | - | 2 - 3 | 198 | 65 |