The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161841.1 | NP_001155313.1 | 15 - 16 | 2725 | 61 |
NM_018837.3 | NP_061325.1 | 15 - 16 | 2386 | 61 | |
NM_198596.2 | NP_940998.2 | 15 - 16 | 2725 | 61 | |
XM_005260458.2 | XP_005260515.1 | 15 - 16 | 2301 | 61 | |
XM_006723827.1 | XP_006723890.1 | 15 - 16 | 2304 | 61 | |
XM_006723828.1 | XP_006723891.1 | 15 - 16 | 2741 | 61 | |
XM_006723829.2 | XP_006723892.1 | 15 - 16 | 2954 | 61 | |
XM_006723830.1 | XP_006723893.1 | 15 - 16 | 2301 | 61 | |
XM_011528913.1 | XP_011527215.1 | 15 - 16 | 2371 | 61 | |
XM_011528914.1 | XP_011527216.1 | 15 - 16 | 2304 | 61 | |
XM_011528915.1 | XP_011527217.1 | 15 - 16 | 2304 | 61 | |
XM_011528916.1 | XP_011527218.1 | 15 - 16 | 2738 | 61 | |
XM_017027955.1 | XP_016883444.1 | 15 - 16 | 2373 | 61 | |
GenBank mRNA | AB033073.2 | - | 15 - 16 | 2386 | 61 |
AK074483.1 | - | 9 - 10 | 1048 | 61 | |
AK075035.1 | - | 8 - 9 | 931 | 61 | |
AL133001.1 | - | 4 - 5 | 478 | 61 | |
AY101176.1 | - | 15 - 16 | 2725 | 61 | |
AY358461.1 | - | 15 - 16 | 2663 | 61 | |
BC020962.1 | - | 4 - 5 | 363 | 61 | |
BC073790.1 | - | 2 - 3 | 120 | 61 | |
BC090952.1 | - | 4 - 5 | 298 | 61 | |
BC110539.1 | - | 14 - 15 | 2152 | 61 | |
CR749319.1 | - | 15 - 16 | 2669 | 61 |