The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037666.2 | NP_001032755.1 | 5 - 6 | 762 | 73 |
GenBank mRNA | AK055587.1 | - | 5 - 6 | 668 | 73 |
BC021927.1 | - | 5 - 6 | 648 | 73 | |
CR456449.1 | - | 5 - 6 | 677 | 73 |