Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
114571
Gene Name
solute carrier family 22 member 9
Gene Aliases
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location
Chr.11: 63368871 - 63410878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017017160.1 XP_016872649.1 2 - 3 511 61
GenBank mRNA BC022379.1 - 2 - 3 779 61

View Full Product Details