Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ST6GALNAC1
Entrez Gene ID
55808
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
HSY11339, SIAT7A, ST6GalNAcI, STYI
Chromosome Location
Chr.17: 76617769 - 76643838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289107.1 NP_001276036.1 5 - 6 1509 73
NM_018414.4 NP_060884.1 4 - 5 1372 73
NR_110309.1 4 - 5 1372 73
XM_011524995.2 XP_011523297.1 4 - 5 1292 73
XM_011524996.2 XP_011523298.1 4 - 5 1292 73
XM_017024842.1 XP_016880331.1 4 - 5 1292 73
XM_017024843.1 XP_016880332.1 4 - 5 1292 73
XM_017024844.1 XP_016880333.1 5 - 6 1392 73
GenBank mRNA AK000113.1 - 3 - 4 526 73
AY096001.1 - 4 - 5 1172 73
AY358918.1 - 4 - 5 1317 73
BC022462.1 - 4 - 5 1227 73
Y11339.2 - 4 - 5 1232 73

View Full Product Details