Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC3B
Entrez Gene ID
154661
Gene Name
RUN domain containing 3B
Gene Aliases
RPIB9, RPIP9
Chromosome Location
Chr.7: 87627656 - 87832298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134405.1 NP_001127877.1 1 - 2 535 72
NM_001134406.1 NP_001127878.1 1 - 2 535 72
NM_138290.2 NP_612147.1 1 - 2 535 72
XM_005250156.2 XP_005250213.1 1 - 2 535 72
XM_011515826.1 XP_011514128.1 1 - 2 535 72
XM_011515827.1 XP_011514129.1 1 - 2 535 72
XM_017011761.1 XP_016867250.1 1 - 2 144 72
XM_017011766.1 XP_016867255.1 1 - 2 335 72
GenBank mRNA AK055233.1 - 1 - 2 533 72
AK293607.1 - 1 - 2 351 72
AK294039.1 - 1 - 2 333 72
BC022520.1 - 1 - 2 486 72
DC304423.1 - 1 - 2 351 72

View Full Product Details