The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281710.1 | NP_001268639.1 | 12 - 13 | 1642 | 119 |
NM_001281711.1 | NP_001268640.1 | 12 - 13 | 1603 | 119 | |
NM_001281712.1 | NP_001268641.1 | 11 - 12 | 1422 | 119 | |
NM_015341.4 | NP_056156.2 | 12 - 13 | 1675 | 119 | |
XM_005263908.3 | XP_005263965.1 | 12 - 13 | 1615 | 119 | |
XM_006712388.3 | XP_006712451.1 | 12 - 13 | 1615 | 119 | |
GenBank mRNA | AK291053.1 | - | 12 - 13 | 1616 | 119 |
AK299248.1 | - | 12 - 13 | 1544 | 119 | |
AK303725.1 | - | 11 - 12 | 1367 | 119 | |
AK308050.1 | - | 12 - 13 | 1582 | 119 | |
BC024211.2 | - | 12 - 13 | 1629 | 119 | |
D38553.1 | - | 12 - 13 | 1605 | 119 |