The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258390.1 | NP_001245319.1 | 6 - 7 | 1019 | 112 |
NM_001258391.1 | NP_001245320.1 | 4 - 5 | 837 | 112 | |
NM_001318799.1 | NP_001305728.1 | 5 - 6 | 629 | 112 | |
NM_178834.4 | NP_849156.1 | 5 - 6 | 995 | 112 | |
XM_006718769.3 | XP_006718832.1 | 5 - 6 | 608 | 112 | |
XM_006718770.3 | XP_006718833.1 | 4 - 5 | 665 | 112 | |
GenBank mRNA | AK055539.1 | - | 5 - 6 | 996 | 112 |
AK293317.1 | - | 5 - 6 | 826 | 112 | |
AK296231.1 | - | 4 - 5 | 625 | 112 | |
AK298165.1 | - | 6 - 7 | 654 | 112 | |
AY358554.1 | - | 6 - 7 | 844 | 112 | |
BC025407.1 | - | 5 - 6 | 788 | 112 |