The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199777.1 | NP_001186706.1 | 9 - 10 | 1463 | 104 |
NM_172240.2 | NP_758440.1 | 10 - 11 | 1482 | 104 | |
NR_037659.1 | 10 - 11 | 1333 | 104 | ||
NR_037660.1 | 8 - 9 | 1291 | 104 | ||
GenBank mRNA | AB053266.1 | - | 9 - 10 | 972 | 104 |
AK027829.1 | - | 6 - 7 | 681 | 104 | |
AK074772.1 | - | 10 - 11 | 1220 | 104 | |
AK123106.1 | - | 9 - 10 | 1460 | 104 | |
AK123625.1 | - | 6 - 7 | 680 | 104 | |
AK295596.1 | - | 10 - 11 | 1071 | 104 | |
AL832918.1 | - | 9 - 10 | 969 | 104 | |
BC026080.2 | - | 10 - 11 | 1244 | 104 | |
EF011619.1 | - | 10 - 11 | 1244 | 104 |