The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243313.1 | NP_001230242.1 | 13 - 14 | 1936 | 77 |
NM_015690.4 | NP_056505.2 | 13 - 14 | 1936 | 77 | |
XM_005246464.1 | XP_005246521.1 | 13 - 14 | 1824 | 77 | |
XM_011510959.1 | XP_011509261.1 | 13 - 14 | 1909 | 77 | |
XM_011510960.1 | XP_011509262.1 | 13 - 14 | 1877 | 77 | |
XM_011510961.1 | XP_011509263.1 | 4 - 5 | 507 | 77 | |
XM_017003804.1 | XP_016859293.1 | 13 - 14 | 1824 | 77 | |
XM_017003805.1 | XP_016859294.1 | 12 - 13 | 1613 | 77 | |
GenBank mRNA | AB033104.1 | - | 13 - 14 | 1893 | 77 |
AF200815.1 | - | 12 - 13 | 1657 | 77 | |
BC026158.2 | - | 13 - 14 | 1876 | 77 |