The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278629.1 | NP_001265558.1 | 8 - 9 | 1211 | 86 |
NM_001278630.1 | NP_001265559.1 | 8 - 9 | 1068 | 86 | |
NM_152786.2 | NP_689999.1 | 8 - 9 | 1284 | 86 | |
XM_005251885.3 | XP_005251942.1 | 6 - 7 | 590 | 86 | |
XM_011518477.1 | XP_011516779.1 | 8 - 9 | 1211 | 86 | |
XM_011518478.1 | XP_011516780.1 | 8 - 9 | 1211 | 86 | |
XM_011518479.2 | XP_011516781.1 | 6 - 7 | 866 | 86 | |
GenBank mRNA | AK313867.1 | - | 8 - 9 | 1200 | 86 |
BC026884.1 | - | 8 - 9 | 1250 | 86 | |
BC033037.2 | - | 8 - 9 | 1169 | 86 |