Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 11 - 12 1435 75
NM_001278630.1 NP_001265559.1 11 - 12 1292 75
NM_152786.2 NP_689999.1 11 - 12 1508 75
XM_005251885.3 XP_005251942.1 9 - 10 814 75
XM_011518477.1 XP_011516779.1 11 - 12 1435 75
XM_011518478.1 XP_011516780.1 11 - 12 1435 75
XM_011518479.2 XP_011516781.1 9 - 10 1090 75
GenBank mRNA AK301872.1 - 11 - 12 1295 75
AK313867.1 - 11 - 12 1424 75
BC026884.1 - 11 - 12 1474 75
BC033037.2 - 11 - 12 1393 75

View Full Product Details