Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 12 - 13 1515 82
NM_001278630.1 NP_001265559.1 12 - 13 1372 82
NM_152786.2 NP_689999.1 12 - 13 1588 82
XM_005251885.3 XP_005251942.1 10 - 11 894 82
XM_011518479.2 XP_011516781.1 10 - 11 1170 82
GenBank mRNA AK313867.1 - 12 - 13 1504 82
BC026884.1 - 12 - 13 1554 82
BC033037.2 - 12 - 13 1473 82

View Full Product Details