Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.19: 40292743 - 40348527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256440.1 NP_001243369.1 5 - 6 429 81
NM_001256441.1 NP_001243370.1 5 - 6 424 81
NR_046202.1 5 - 6 477 81
XM_005258520.2 XP_005258577.1 5 - 6 421 81
XM_011526460.2 XP_011524762.1 5 - 6 417 81
XM_017026291.1 XP_016881780.1 5 - 6 432 81
XM_017026292.1 XP_016881781.1 4 - 5 338 81
XM_017026293.1 XP_016881782.1 5 - 6 432 81
XM_017026294.1 XP_016881783.1 2 - 3 147 81
GenBank mRNA AK094207.1 - 5 - 6 417 81
AL834131.1 - 5 - 6 454 81
BC027935.1 - 5 - 6 445 81
BI916290.1 - 5 - 6 424 81
BM459040.1 - 5 - 6 374 81
CR936760.1 - 5 - 6 430 81

View Full Product Details